Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals LY 294002, 98%

Catalog No. AAJ67289LB0
Click to view available options
Quantity:
5 mg
10 mg
25 mg

Catalog No. AAJ67289LB0

Supplier: Thermo Scientific Chemicals J67289LB0

Only null left
Add to Cart

CAS: 154447-36-6 | C19H17NO3, C19H17NO3 | 307.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154447-36-6
Molecular Formula C19H17NO3
Synonym 2-(4-morpholinyl)-8-phenyl-4H-1-benzopyran-4-one;

Specifications

Quantity 5 mg
Description White to pale yellow
Content And Storage -30°C to -10°C
Formula Weight 307.35g/mol
Form solid
InChI Key CZQHHVNHHHRRDU-UHFFFAOYSA-N
SMILES O=C1C=C(OC2=C(C=CC=C12)C1=CC=CC=C1)N1CCOCC1
IUPAC Name 2-(morpholin-4-yl)-8-phenyl-4H-chromen-4-one
Molecular Weight (g/mol) 307.35
Assay 98%
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.