Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 294002 hydrochloride
GREENER_CHOICE

Catalog No. 113025
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Prototypical PI 3-kinase inhibitor; also inhibits other kinases

LY 294002 hydrochloride is a prototypical PI 3-kinase inhibitor (IC50 values are 0.31, 0.73, 1.06 and 6.60 μM for PI 3-Kβ, PI 3-Kα, PI 3-Kδ and PI 3-Kγ respectively). Also inhibits other kinases including, CK2, mTOR, PLK1, PIM1 and PIM3. Inhibits proliferation and induces apoptosis in human colon cancer cells in vitro and in vivo. Suppresses proliferation of mESCs. Inhibits autophagic sequestration in rat hepatocytes.

Negative Control also available.

Chemical Identifiers

CAS 934389-88-5
Molecular Formula C19H18ClNO3

Specifications

Quantity 25 mg
Formula Weight 343.81
Purity >99%
Molecular Weight (g/mol) 343.807
Chemical Name or Material LY 294002 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.