Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 2955303

Catalog No. p-7166772
Click to view available options
Quantity:
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity and selective RARγ antagonist

LY 2955303 is a high affinity and selective RARγ antagonist (Ki values are 1.1 nM, >1.7 μM and >2.9 μM for RARγ, RARα and RARβ respectively). Displays analgesic efficacy in a mouse model of osteoarthritis-like joint pain.

Specifications

Chemical Name or Material 4-[5-[3,5-Bis(1,1-dimethylethyl)phenyl]-1-[4-[(4-methyl-1-piperazinyl)carbonyl]phenyl]-1H-pyrazol-3-yl]benzoic acid
CAS 1433497-19-8
Quantity 5 mg
Solubility Soluble to 20 mM in DMSO
Molecular Formula C36H42N4O3
Formula Weight 578.74
Product Type Antagonist
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.