Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 310762 hydrochloride
GREENER_CHOICE

Catalog No. 307810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-781-0 10 mg
30-785-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-781-0 Supplier Tocris Bioscience™ Supplier No. 3078/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective 5-HT1D antagonist

5-HT1D-preferring receptor antagonist (EC50 = 31 nM). Displays no activity on 5-HT transport.

Chemical Identifiers

CAS 192927-92-7
Molecular Formula C24H28ClFN2O2

Specifications

Quantity 10 mg
Formula Weight 430.94
Purity >98%
Molecular Weight (g/mol) 430.948
Chemical Name or Material LY 310762 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.