Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ LY 341495 disodium salt

Click to view available options
Quantity:
1 mg
10 mg
Description
Highly potent and selective group II metabotropic glutamate receptor antagonist. Disodium salt of LY 341495 (Cat. No. 1209).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 397.33 |
Purity | >97% |
Chemical Name or Material | LY 341495 disodium salt |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction