Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 341495
GREENER_CHOICE

Catalog No. p-4783615
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent and selective group II mGlu antagonist

Highly potent and selective group II metabotropic glutamate receptor antagonist (Ki/IC50 values are 2.3, 1.3, 173, 990, 6800, 8200 and 22000 nM for human mGlu2, mGlu3, mGlu8, mGlu7a, mGlu1a, mGlu5a and mGlu4a receptors respectively). Readily brain penetrant and active in vivo.

Disodium salt (Cat. No. 4062) also available.

Chemical Identifiers

CAS 201943-63-7
Molecular Formula C20H19NO5

Specifications

Quantity 1 mg
Formula Weight 353.37
Purity >98%
Molecular Weight (g/mol) 353.374
Chemical Name or Material LY 341495
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.