Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 379268 disodium salt
GREENER_CHOICE

Catalog No. p-5545874
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium salt of LY 379268 (Cat. No. 2453); selective group II mGlu receptor agonist

LY 379268 disodium salt is a sodium salt of LY 379268 (Cat. No. 2453), a highly selective group II mGlu receptor agonist.

Specifications

Quantity 10 mg
Product Type LY 379268 disodium salt
Purity >98%
Molecular Weight (g/mol) 231.11
Inhibitors LY 379268 disodium salt

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.