Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 379268
GREENER_CHOICESDP

Catalog No. 245310 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-531-0 10 mg
24-535-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-531-0 Supplier Tocris Bioscience™ Supplier No. 2453/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly selective group II mGlu agonist

LY 379268 is a highly selective group II mGlu receptor agonist (EC50 values are 2.69 and 4.48 nM for hmGlu2 and hmGlu3 respectively) that displays > 80-fold selectivity over group I and group III receptors. Provides protection against NMDA-mediated cell death in vitro and offers almost complete protection against CA1 hippocampal damage following global ischemia in gerbils. Orally and systemically active. LY379268 disodium salt also available.

Chemical Identifiers

CAS 191471-52-0
Molecular Formula C7H9NO5

Specifications

Quantity 10 mg
Formula Weight 187.15
Purity >99%
Molecular Weight (g/mol) 187.151
Chemical Name or Material LY 379268
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.