Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ LY 450139

Click to view available options
Quantity:
5 mg
25 mg
Description
LY 450139 is a γ-secretase pseudo-inhibitor. Inhibits Aβ secretion from cells in vitro and increases accumulation of γ-secretase products and long Aβ species in vitro and in vivo. Also attenuates the accumulation of activated microglia and neurological injury in a traumatic brain injury animal model.
Specifications
Specifications
| Chemical Name or Material | (2S)-2-Hydroxy-3-methyl-N-[(1S)-1-methyl-2-oxo-2-[[(1S)-2,3,4,5-tetrahydro-3-methyl-2-oxo-1H-3-benzazepin-1-yl]amino]ethyl]butanamide |
| CAS | 425386-60-3 |
| Quantity | 25 mg |
| Molecular Formula | C19H27N3O4 |
| Formula Weight | 361.44 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction