Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 487379 hydrochloride
GREENER_CHOICE

Catalog No. 328310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-831-0 10 mg
32-835-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-831-0 Supplier Tocris Bioscience™ Supplier No. 3283/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective positive allosteric modulator of mGlu2 receptors

Positive allosteric modulator selective for mGlu2 receptors. Potentiates glutamate-stimulated [35S]GTPγS binding (EC50 values are 1.7 and > 10 μM for mGlu2 and mGlu3 receptors respectively). Devoid of any activity at mGlu5 and mGlu7 receptors. Promotes cognitive flexibility in a rat model.

Chemical Identifiers

CAS 353229-59-1
Molecular Formula C21H20ClF3N2O4S

Specifications

Quantity 10 mg
Formula Weight 488.91
Purity >99%
Molecular Weight (g/mol) 488.906
Chemical Name or Material LY 487379 hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.