Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-49A Mouse anti-Mouse, Brilliant Violet 421, Clone: A1, BD Optibuild™

Catalog No. BDB742364 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742364 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742364 Supplier BD Biosciences Supplier No. 742364
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The A1 monoclonal antibody specifically binds to the Ly-49A[B6] alloantigen, an inhibitory receptor that is expressed on subsets of natural killer (NK) cells and NK-1.1-positive T lymphocytes (NKT cells) in C57BL/6, C57BL/10, and B10 congenic mice, on a population of memory CD8+ T lymphocytes and NK1.1+ T cells in C57BL/6 mice, and on a distinct subset of B-1 cells (CD5+B220[lo]) of C57BL/6 mice. The A1 antibody has also been reported to crossreact with Ly-49ANOD, Ly-49PNOD, Ly-49P129/J, and Ly-49V129/J alloantigens. The proportion of NKT cells expressing Ly-49A is higher in thymus than in liver, and there is evidence that the down regulation of Ly-49 receptor expression is necessary for normal NKT cell development to occur. Most NK cells express a single allele of Ly-49A, although they may express more than one. The Ly-49 family of NK-cell receptors, of the C-type lectin superfamily, are disulfide-linked type-II transmembrane protein homodimers with extracellular carbohydrate-recognition domains (CRD) that bind to MHC class I alloantigens. The A1 antibody is specific for the Ly-49A[B6] CRD. The Ly-49 family members are expressed independently, so an individual NK or T cell may display more than one class of Ly-49 receptor homodimers. The Ly-49A[B6] allonantigen binds to H-2D[d], H-2D[k], and H-2D[p], and the A1 antibody blocks this binding. Binding of Ly-49A[B6] to lyphoblasts expressing MHC class I antigens of the f, q, r, s, and v haplotypes has also been demonstrated.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49A
Classification Monoclonal
Clone A1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ly-49a, Klra1, Killer cell lectin-like receptor subfamily A member1, Klra22
Host Species Mouse
Immunogen Mouse C57BL/6N T lymphoma EL-4
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.