Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-49l Antibody (YLI-90), Biotin, Novus Biologicals™
SDP

Catalog No. NBP128094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128094 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128094 Supplier Novus Biologicals Supplier No. NBP128094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ly-49l Monoclonal specifically detects Ly-49l in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Ly-49l
Applications Flow Cytometry
Classification Monoclonal
Clone YLI-90
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Flow Cytometry <= 1 ug/10^6 cells
Gene Alias Killer cell lectin like receptor subfamily A member 9, Klra9, Ly49I receptor, LY49I1
Gene Symbols Klra9
Host Species Mouse
Immunogen CHO-K1 cells transfected with B6 allele of Ly49I gene
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16640
Test Specificity B6 allele of the Ly49I receptor
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.