Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6A/E Antibody (RM0330-2M39), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP211871FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211871FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211871FR Supplier Novus Biologicals Supplier No. NBP211871FR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6A/E Monoclonal specifically detects Ly-6A/E in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Ly-6A/E
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RM0330-2M39
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Ly 6A/E, Ly6A.2, Ly6A/E, Ly6E.1, Lymphocyte antigen 6 complex locus A, Lymphocyte antigen 6 complex locus E, Retinoic acid induced gene E, RIGE, SCA1, SCA-1, SCA2, Stem cell antigen 1, Stem cell antigen 2, T cell activating protein, TAP, Thymic shared antigen 1, TSA1
Gene Symbols LY6A
Host Species Rat
Immunogen Mouse recombinant Ly-6A/E
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 110454
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.