Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6B.2 Antibody (7/4), PE, Novus Biologicals™
SDP

Catalog No. NBP213077PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213077PE 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213077PE Supplier Novus Biologicals Supplier No. NBP213077PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6B.2 Monoclonal specifically detects Ly-6B.2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Ly-6B.2
Applications Flow Cytometry
Classification Monoclonal
Clone 7/4
Conjugate PE
Dilution Flow Cytometry
Host Species Rat
Immunogen Cultured bone marrow cells
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Ly6B.2, a polymorphic 40 kDa antigen expressed by polymorphonuclear cells, but absent on resident tissue macrophages. Strains positive for 7/4:129J; AKR; C57BL/6; C57BL/10; C58; DBA/2; NZB; NZW; SJL; MFI; Swiss (PO) Strains Negative/Weak for 7/4:A2G; A/Sn; ASW; BALB/c; C3H/HEH: CBA.T6T6.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.