Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6C Antibody (ER-MP20), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB10065413W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065413W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065413W Supplier Novus Biologicals Supplier No. NB10065413AF405
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen Ly-6C
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone ER-MP20
Conjugate Alexa Fluor 405
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C
Gene Symbols Ly6c1
Host Species Rat
Immunogen Balb/c macrophage precursor cell hybrids
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17067
Test Specificity Ly6C (ER-MP20)
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.