Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6C Antibody (ER-MP20), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10065413S Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065413S 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065413S Supplier Novus Biologicals Supplier No. NB10065413R
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen Ly-6C
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone ER-MP20
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C
Gene Symbols LY6C1
Host Species Rat
Immunogen Balb/c macrophage precursor cell hybrids
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 17067
Test Specificity Ly6C (ER-MP20)
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.