Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6C Antibody (HK1.4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP128046FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128046FR 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128046FR Supplier Novus Biologicals Supplier No. NBP128046FR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Ly-6C
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HK1.4
Conjugate DyLight 680
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Alias Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C
Gene Symbols LY6C1
Host Species Rat
Immunogen L3 cloned CTL cells
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17067
Test Specificity Murine Ly-6C, Mr 18 kDa
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2c
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.