Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6G/Ly-6C Antibody (RM0064-6D17), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP21784646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP21784646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP21784646 Supplier Novus Biologicals Supplier No. NBP211784JF646
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ly-6G/Ly-6C
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0064-6D17
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GR, Gr-1, Ly-6, Ly6G
Gene Symbols LY6G
Host Species Rat
Immunogen Cell lysates of mouse neutrophils Ig
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 546644
Test Specificity This antibody detects mouse Ly-6G (Gr-1), the LY-6G/LY-6C complex.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.