Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G/Ly-6C Rat anti-Mouse, Clone: RB6-8C5, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP3119790.2MG
Description
Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
Ly-6G/Ly-6C | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
GR, Gr-1, Ly-6, Ly6G | |
This antibody was raised in Lou-M rats using a popliteal lymph node immunisation and fusion protocol | |
0.2 mg | |
Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell | |
546644 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
RB6-8C5 | |
Unconjugated | |
PBS | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction