Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G/Ly-6C Rat anti-Mouse, Clone: RB6-8C5, Novus Biologicals™

Rat Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | Ly-6G/Ly-6C |
---|---|
Clone | RB6-8C5 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
Description
Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
Ly-6G/Ly-6C | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
RUO | |
Mouse | |
GR, Gr-1, Ly-6, Ly6G | |
This antibody was raised in Lou-M rats using a popliteal lymph node immunisation and fusion protocol | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
RB6-8C5 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Monoclonal | |
Rat | |
Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell | |
PBS | |
546644 | |
IgG2a κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title