Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6G6F Antibody, Novus Biologicals™
SDP

Catalog No. NBP224668SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224668SS 0.025 mg
NBP224668 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224668SS Supplier Novus Biologicals Supplier No. NBP224668SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ly-6G6F Polyclonal specifically detects Ly-6G6F in Human, Goat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ly-6G6F
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0-3.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q5SQ64
Gene Alias C6orf21, G6F, locus G6D, LY6G6D, lymphocyte antigen 6 complex locus protein G6f, lymphocyte antigen 6 complex, locus G6F, NG32
Gene Symbols LY6G6F
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 66-100 of human G6D was used as immunogen, GenBank no NP_001003693.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 259215
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 95% homologous in mouse, horse, rat, rhesus monkey and dog, 90% homologous in cow, 80% homologous in platypus and 70% homologous in opossum.
Target Species Human, Goat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.