Learn More
Invitrogen™ Ly-6K Monoclonal Antibody (B33fc8k), PE, eBioscience™
Rat Monoclonal Antibody
$318.00
Specifications
Antigen | Ly-6K |
---|---|
Clone | B33fc8k |
Concentration | 0.2 mg/mL |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-2357
![]() |
Invitrogen™
12592982 |
100 μg |
Each of 1 for $318.00
|
|
|||||
Description
Description: This B33fc8k monoclonal antibody reacts with mouse Ly-6k, a member of the Ly6/uPAR superfamily. Ly-6k is a GPI linked protein predominantly found on the cell surface. One study suggests expression may also be non-membranous. Expression is found on B cells and reported to be on plasma cells as well as cells in the testis. Using the B33fc8k monoclonal antibody, expression is on a B lymphocyte cell subset in the spleen that correlates to transitional cells (CD23-IgM-). In humans Ly-6k has been found associated with carcinomas of the head and testis. Applications Reported: This B33fc8k antibody has been reported for use in flow cytometric analysis. Applications Tested: This B33fc8k antibody has been tested by flow cytometric analyis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Required for sperm migration into the oviduct and male fertility by controlling binding of sperm to zona pellucida. May play a role in cell growth (PubMed:18089789). [UniProt].Specifications
Ly-6K | |
0.2 mg/mL | |
Flow Cytometry | |
PE | |
Rat | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9CWP4 | |
76486 | |
Primary | |
Affinity chromatography |
B33fc8k | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Mouse | |
LY6K | |
2410015A16Rik; 3110035B01Rik; cancer/testis antigen 97; CO16; CT97; FLJ35226; HSJ001348; Ly6k; ly-6K; lymphocyte antigen 6 complex, locus K; lymphocyte antigen 6 complex, locus K-like; lymphocyte antigen 6K; mLy-6K; up-regulated in lung cancer 10; URLC10 | |
LY6K | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.