Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly49A Mouse anti-Mouse, Unconjugated, Clone: A1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF2090100UG
Description
This mouse monoclonal Anti-Ly49A, clone A1, Cat. No. MABF2090 is tested for use in Flow Cytometry, Immunocytochemistry and Immunoprecipitation, for the detection of Lymphocyte antigen 49a.
Specifications
Ly49A | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Klra1;LY-49;Ly-49a;LY49A | |
EL-4 T lymphoma cells. | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Antibodies | |
IgG2a κ |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
A1 | |
Unconjugated | |
T-cell surface glycoprotein YE1/48;Lymphocyte antigen 49a;Ly-49a;T lymphocyte antigen A1 | |
Mouse | |
30.50 kDa calculated. | |
100 μg | |
Inflammation & Immunology | |
Clone A1 specifically detects Lymphocyte antigen 49a (LY49A) in murine species. | |
Stable for 1 year at 2-8°C from date of receipt. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction