Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly49A Mouse anti-Mouse, Unconjugated, Clone: A1, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $456.56
Specifications
Antigen | Ly49A |
---|---|
Clone | A1 |
Concentration | Please refer to lot specific datasheet. |
Applications | Flow Cytometry, Immunocytochemistry, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF2090-25
|
MilliporeSigma
MABF209025UG |
25 μg |
Each for $192.35
|
|
|||||
MABF2090-100
|
MilliporeSigma
MABF2090100UG |
100 μg |
Each for $456.56
|
|
|||||
Description
This mouse monoclonal Anti-Ly49A, clone A1, Cat. No. MABF2090 is tested for use in Flow Cytometry, Immunocytochemistry and Immunoprecipitation, for the detection of Lymphocyte antigen 49a.Specifications
Ly49A | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
T-cell surface glycoprotein YE1/48;Lymphocyte antigen 49a;Ly-49a;T lymphocyte antigen A1 | |
EL-4 T lymphoma cells. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
A1 | |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Inflammation & Immunology | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Klra1;LY-49;Ly-49a;LY49A | |
IgG2a κ | |
Protein G purified | |
Clone A1 specifically detects Lymphocyte antigen 49a (LY49A) in murine species. | |
30.50 kDa calculated. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title