Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly49C Antibody (14B11) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP128082 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128082 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128082 Supplier Novus Biologicals Supplier No. NBP128082
Add to Cart
Add to Cart

Golden Syrian Hamster Monoclonal Antibody

Ly49C Monoclonal specifically detects Ly49C in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Ly49C
Applications Flow Cytometry
Classification Monoclonal
Clone 14B11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry <= 1 ug/10^6 cells
Gene Alias 5E6, Cmv1, Killer cell lectin like receptor 3, Killer cell lectin like receptor 6, Killer cell lectin like receptor 8, Killer cell lectin like receptor 9, Klra3, Klra6, Klra8, Klra9, Ly49 C antigen, Ly49 F antigen, Ly49 H antigen, Ly49 I antigen, Lymphocyte antigen 49C, Lymphocyte antigen 49F, Lymphocyte antigen 49H, Lymphocyte antigen 49I, T cell surface glycoprotein Ly 49C, T cell surface glycoprotein Ly 49F, T cell surface glycoprotein Ly 49H, T cell surface glycoprotein Ly 49I
Host Species Golden Syrian Hamster
Immunogen IL-2 activated natural killed (A-LAK) cells from C57BL/6 mice
Purification Method Protein A or G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity An epitope common to the Ly49C, Ly49F, Ly49H and Ly49I allelic/splice variants
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.