Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly49F Antibody (HBF-719), Biotin, Novus Biologicals™
SDP

Catalog No. NBP128098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128098 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128098 Supplier Novus Biologicals Supplier No. NBP128098
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ly49F Monoclonal specifically detects Ly49F in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Ly49F
Applications Flow Cytometry
Classification Monoclonal
Clone HBF-719
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Flow Cytometry <= 1 ug/10^6 cells
Gene Alias Killer cell lectin like receptor 6, Klra6, Ly49F antigen, Lymphocyte antigen 49F, T cell surface glycoprotein Ly 49F
Gene Symbols Klra6
Host Species Mouse
Immunogen CHO-K1 cells transfected with B6 allele of Ly49F gene
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16637
Test Specificity B6 allele of the Ly49F receptor
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.