Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LY6G5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570790 Supplier Invitrogen™ Supplier No. PA570790
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 85%; Human: 100%; Rabbit: 77%; Rat: 77%.

LY6G5C belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LY6G5C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ly6g5c
Gene Accession No. Q5SRR4
Gene Alias C6orf20; Canine epididymal gene product 8; CE8; CE8 protein, variant 2; epididymal secretory protein 8; G5C; Ly6g5c; LY6G5CA; LY6G5CB; Lymphocyte antigen 6 complex locus protein G5c; lymphocyte antigen 6 complex, locus G5C; lymphocyte antigen 6 family member G5C; lymphocyte antigen-6 G5C; NG33; re8
Gene Symbols Ly6g5c
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human LY6G5C.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80741
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.