Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly6K Antibody (750747) [DyLight 650], Novus Biologicals™
SDP

Catalog No. NB376036 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376036 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376036 Supplier Novus Biologicals Supplier No. FAB6648W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ly6K Monoclonal antibody specifically detects Ly6K in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Ly6K
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 750747
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Ly6K, Met1-Gly138, Accession # Q17RY6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54742
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.