Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly6K Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224405 0.1 mg
NB019387 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224405 Supplier Novus Biologicals Supplier No. NBP224405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ly6K Polyclonal specifically detects Ly6K in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Ly6K
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K
Gene Symbols LY6K
Host Species Rabbit
Immunogen Reacts with residues 82-100 RQRDPEDSQRTDEGDNRVW of the 14 kDa human Ly-6K protein.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54742
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.