Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly6K Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | Ly6K |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Description
Ly6K Polyclonal specifically detects Ly6K in Human samples. It is validated for Western Blot, ELISA.Specifications
Ly6K | |
Western Blot, ELISA 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Human | |
cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K | |
LY6K | |
IgG | |
Affinity Purified |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
54742 | |
Reacts with residues 82-100 RQRDPEDSQRTDEGDNRVW of the 14 kDa human Ly-6K protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title