Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYAG/GAA Antibody (5E5), Novus Biologicals™
SDP

Catalog No. NB297852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297852 50 μL
NB297853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297852 Supplier Novus Biologicals Supplier No. NBP32656250UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LYAG/GAA Monoclonal antibody specifically detects LYAG/GAA in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen LYAG/GAA
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5E5
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Acid maltase, Aglucosidase alfa, EC 3.2.1.20, glucosidase, alpha, acid, LYAG, lysosomal alpha-glucosidase
Host Species Rabbit
Immunogen A synthesized peptide derived from Human LYAG/GAA [UniProt P10253]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2548
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.