Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYAG/GAA Antibody (5E5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | LYAG/GAA |
---|---|
Clone | 5E5 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
LYAG/GAA Monoclonal antibody specifically detects LYAG/GAA in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
LYAG/GAA | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Acid maltase, Aglucosidase alfa, EC 3.2.1.20, glucosidase, alpha, acid, LYAG, lysosomal alpha-glucosidase | |
A synthesized peptide derived from Human LYAG/GAA [UniProt P10253] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5E5 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2548 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title