Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYAR Antibody, Novus Biologicals™
SDP

Catalog No. NBP15647420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15647420 20 μL
NBP156474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15647420 Supplier Novus Biologicals Supplier No. NBP15647420UL

Rabbit Polyclonal Antibody

LYAR Polyclonal specifically detects LYAR in Human samples. It is validated for Western Blot.

Specifications

Antigen LYAR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NX58
Gene Alias cell growth-regulating nucleolar protein, FLJ20425, Ly1 reactive homolog (mouse), ZLYAR
Gene Symbols LYAR
Host Species Rabbit
Immunogen Synthetic peptides corresponding to LYAR(Ly1 antibody reactive homolog (mouse)) The peptide sequence was selected from the middle region of LYAR. Peptide sequence EQQGEVKKNKRERKEERQKKRKREKKELKLENHQENSRNQKPKKRKKGQE.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 55646
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.