Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYK5 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB326034 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326034 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326034 Supplier Novus Biologicals Supplier No. NBP299336MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LYK5 Polyclonal antibody specifically detects LYK5 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen LYK5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias LYK5FLJ90524, NY-BR-96, protein kinase LYK5, Serologically defined breast cancer antigen NY-BR-96, STE20-like pseudokinase, STE20-related adapter protein, STE20-related adaptor protein, STE20-related kinase adapter protein alpha, STE20-related kinase adaptor alpha, Stlk, STRAD alpha, STRADPMSE
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human LYK5.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 92335
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.