Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lyl1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227188 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227188 0.1 mg
NBP227188SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227188 Supplier Novus Biologicals Supplier No. NBP227188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lyl1 Polyclonal specifically detects Lyl1 in Canine, Human, Pig, Primate samples. It is validated for Simple Western, Western Blot.

Specifications

Antigen Lyl1
Applications Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml, Simple Western 1:10
Formulation No buffer with No Preservative
Gene Alias BHLHA18, bHLHa18Class A basic helix-loop-helix protein 18, lymphoblastic leukemia derived sequence 1, Lymphoblastic leukemia-derived sequence 1, protein lyl-1
Gene Symbols LYL1
Host Species Rabbit
Immunogen 235 RSQPAPPADPDGSPGG 250
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4066
Test Specificity Dog, Human, Orangutan, Wild pig.
Target Species Canine, Human, Pig, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.