Learn More
Description
Specifications
Specifications
| Antigen | Lyl1 |
| Applications | Simple Western, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 3-5ug/ml, Simple Western 1:10 |
| Formulation | No buffer with No Preservative |
| Gene Alias | BHLHA18, bHLHa18Class A basic helix-loop-helix protein 18, lymphoblastic leukemia derived sequence 1, Lymphoblastic leukemia-derived sequence 1, protein lyl-1 |
| Gene Symbols | LYL1 |
| Host Species | Rabbit |
| Immunogen | 235 RSQPAPPADPDGSPGG 250 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
