Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lymphotoxin beta R/TNFRSF3 Antibody (157108), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB315409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315409 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315409 Supplier Novus Biologicals Supplier No. FAB10081PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Lymphotoxin beta R/TNFRSF3
Applications Flow Cytometry
Classification Monoclonal
Clone 157108
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Lymphotoxin beta R/TNFRSF3, Ser28-Pro218, Accession # P50284
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 4055
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.