Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lymphotoxin beta R/TNFRSF3 Antibody (OTI3G10), HRP, Novus Biologicals™
SDP

Catalog No. NBP272551H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272551H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272551H Supplier Novus Biologicals Supplier No. NBP272551H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Lymphotoxin beta R/TNFRSF3
Applications Western Blot
Classification Monoclonal
Clone OTI3G10
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Gene Symbols LTBR
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 31-227 of human Lymphotoxin beta R/TNFRSF3 (NP_002333) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 4055.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.