Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lymphotoxin beta R/TNFRSF3 Rat anti-Human, APC, Clone: 6F4, Novus Biologicals™
SDP

Catalog No. NB173069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173069 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173069 Supplier Novus Biologicals Supplier No. NBP280080APC
This item is not returnable. View return policy

Rat Monoclonal Antibody

Lymphotoxin beta R/TNFRSF3 Monoclonal antibody specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Lymphotoxin beta R/TNFRSF3
Applications Flow Cytometry
Classification Monoclonal
Clone 6F4
Conjugate APC
Formulation PBS
Gene Alias CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Host Species Rat
Immunogen Ectodomain of human Lymphotoxin beta R/TNFRSF3 fused to human IgG Fc.
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 4055
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.