Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lyn Antibody (LYN-01) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500519 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500519 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500519 Supplier Novus Biologicals Supplier No. NB500519
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lyn Monoclonal specifically detects Lyn in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Lyn
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone LYN-01
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:50
Gene Accession No. P07948
Gene Alias EC 2.7.10, EC 2.7.10.2, FLJ26625, JTK8, tyrosine-protein kinase Lyn, v-yes-1 Yamaguchi sarcoma viral related oncogene homolog, Yamaguchi sarcoma viral (v-yes-1) related oncogene homolog
Gene Symbols LYN
Host Species Mouse
Immunogen Bacterially expressed recombinant fragment of human Lyn, aa 8 - 238 (NM_001111097.2).
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4067
Test Specificity The LYN-01 reacts with Lyn (p56/p53), a non-receptor Src-family tyrosine kinase expressed in hematopoietic tissues.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.