Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYPD8 Antibody (961703), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345865 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345865 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345865 Supplier Novus Biologicals Supplier No. FAB9087MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LYPD8 Monoclonal specifically detects LYPD8 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen LYPD8
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 961703
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with human LYPD8, Met1-Asn215, Accession # Q6UX82
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 646627
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.