Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYPLA2 Antibody (02), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB328523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328523 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328523 Supplier Novus Biologicals Supplier No. NBP305819MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LYPLA2 Monoclonal antibody specifically detects LYPLA2 in Human samples. It is validated for ELISA

Specifications

Antigen LYPLA2
Applications ELISA
Classification Monoclonal
Clone 02
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias acyl-protein thioesterase 2, APT2, DJ886K2.4, EC 3.1.2, EC 3.1.2.-, LPL-II, lysophospholipase IIAPT-2LysoPLA II
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LYPLA2 (Uniprot#: O95372; Met 1-Val 231).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11313
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.