Learn More
Description
Specifications
Specifications
| Antigen | Lys20p and Lys21p |
| Applications | Immunohistochemistry, Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 31F5 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:10000-1:50000, Immunohistochemistry 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:1000-1:10000 |
| Formulation | Tissue culture supernatant with 5mM Sodium Azide |
| Gene Alias | LYS20 Lys20p |
| Host Species | Mouse |
| Immunogen | Yeast nuclear preparation |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
