Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lys20p and Lys21p Antibody (31F5), Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | Lys20p and Lys21p |
|---|---|
| Clone | 31F5 |
| Dilution | Western Blot 1:10000-1:50000, Immunohistochemistry 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:1000-1:10000 |
| Applications | Immunohistochemistry, Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Monoclonal |
Description
Lys20p and Lys21p Monoclonal specifically detects Lys20p and Lys21p in Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Lys20p and Lys21p | |
| Western Blot 1:10000-1:50000, Immunohistochemistry 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:1000-1:10000 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| LYS20 Lys20p | |
| Yeast nuclear preparation | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 47.1/48.6 kDa |
| 31F5 | |
| Immunohistochemistry, Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Yeast | |
| 851346 | |
| IgG1 | |
| Tissue culture supernatant | |
| Lys20p and Lys21p |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title