Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine Hydroxylase 2/PLOD2 Antibody (OTI6D1), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP2726A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2726A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2726A30 Supplier Novus Biologicals Supplier No. NBP272556AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysine Hydroxylase 2/PLOD2 Monoclonal specifically detects Lysine Hydroxylase 2/PLOD2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Lysine Hydroxylase 2/PLOD2
Applications Western Blot
Classification Monoclonal
Clone OTI6D1
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 2-oxoglutarate 5-dioxygenase (lysine hydroxylase) 2, 2-oxoglutarate 5-dioxygenase 2, procollagen-lysine, telopeptide lysyl hydroxylase
Gene Symbols PLOD2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 359-640 of human Lysine Hydroxylase 2/PLOD2 (NP_891988) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5352
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.