Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB107015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107015 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107015 Supplier Novus Biologicals Supplier No. NBP298966AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Polyclonal specifically detects Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias EC 1.14.11, EC 1.14.11.-, JHDM2AJMJD1AJHMD2A, JmjC domain-containing histone demethylation protein 2A, JMJD1, jumonji C domain-containing histone demethylase 2A, jumonji domain containing 1, jumonji domain containing 1A, Jumonji domain-containing protein 1A, KIAA0742DKFZp686A24246, lysine (K)-specific demethylase 3A, lysine-specific demethylase 3A, testis-specific protein A, TSGADKFZp686P07111
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 55818
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.