Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB338281 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338281 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338281 Supplier Novus Biologicals Supplier No. NBP149602PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 1.14.11, EC 1.14.11.-, JHDM3AJMJD2, JmjC domain-containing histone demethylation protein 3A, JMJD2Alysine-specific demethylase 4A, jumonji domain containing 2, jumonji domain containing 2A, Jumonji domain-containing protein 2A, KIAA0677jumonji C domain-containing histone demethylase 3A, lysine (K)-specific demethylase 4A
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human JMJD2A protein (within residues 320-460). [Swiss-Prot O75164]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 9682
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.