Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP149600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149600 0.1 mL
NBP149600SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP149600 Supplier Novus Biologicals Supplier No. NBP149600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Polyclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:10000, Simple Western 1:12.5, Chromatin Immunoprecipitation reported in scientific literature (PMID 23884959; 23129632), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23129632), Immunohistochemistry-Paraffin 1:100, Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9H3R0, Q8VCD7
Gene Alias EC 1.14.11, EC 1.14.11.-, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, lysine-specific demethylase 4C
Gene Symbols KDM4C
Host Species Rabbit
Immunogen A partial recombinant protein made to an internal region of the human JMJD2C protein (within residues 450-600). [Swiss-Prot Q9H3R0]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23081
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.