Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB11038884R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11038884R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11038884R Supplier Novus Biologicals Supplier No. NB11038884R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Polyclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.14.11, EC 1.14.11.-, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, lysine-specific demethylase 4C
Gene Symbols KDM4C
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 475 and 525 of human Jumonji Domain Containing 2C using the numbering given in entry NP_055876.2
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23081
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.