Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Antibody (OTI5B9), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N272553J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272553J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272553J646 Supplier Novus Biologicals Supplier No. NBP272553JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Monoclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5B9
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.14.11, EC 1.14.11.-, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, lysine-specific demethylase 4C
Gene Symbols KDM4C
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Chromatin Modifiers, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23081.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.